missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Cyclopentene-1-carboxylic acid, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC396830050

 View more versions of this product

Catalog No. AC396830050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

112.13
100.0
C6H8O2
C5H7COOH
97.5
Glass bottle
5g
1560-11-8
98%
1-Cyclopentene-1-carboxylic acid
MFCD00001396

Chemical Identifiers

1560-11-8
100.0
C6H8O2
112.13
1-Cyclopentene-1-carboxylic acid
97.5
98%
C5H7COOH
MFCD00001396

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.