Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Cyclohexylpiperidine, 97%

$42.50 - $545.45

Chemical Identifiers

Chemical Name or Material 1-Cyclohexylpiperidine
CAS 3319-01-5
Molecular Formula C11H21N
Formula Weight 167.3
MDL Number MFCD00014642
Beilstein 105594
Synonym 1-Piperidinylcyclohexane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0789709

  SDS
alfa aesar™
L0789709
10g Each for $42.50

AAL0789730

  SDS
alfa aesar™
L0789730
250g Each for $545.45
Description & Specifications

Specifications

Formula Weight 167.3
Chemical Name or Material 1-Cyclohexylpiperidine
MDL Number MFCD00014642
CAS 3319-01-5
Assay 97%
Health Hazard 1 Warning
Molecular Formula C11H21N
Boiling Point 231° to 234°C
RTECS Number TM6520000
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 1-Piperidinylcyclohexane
Quantity 10g
Beilstein 105594
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Applications
1-Cyclohexylpiperidine is used in the preparation of 1-(1-phenylcyclohexyl)piperidine, which shows anticholinesterase activity and antagonistic activity to acetylcholine. It is used as a N-methyl-D-aspartate receptor (NMDAR) antagonist. Further, it is used as an anesthetic for animals.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.