missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-(Cyclohexylmethyl)piperazine, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H5549106
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1-(Cyclohexylmethyl)piperazine | |
Colorless | |
230°C to 231°C | |
C11H22N2 | |
MFCD01075237 | |
1-cyclohexylmethyl piperazine, piperazine, 1-cyclohexylmethyl, cyclohexylmethyl piperazine, 1-cyclohexylmethyl-piperazine, 1-cyclohexyl-methyl piperazine, piperazine,1-cyclohexylmethyl | |
C(C1CCCCC1)N1CCNCC1 | |
182.31 | |
182.31 | |
Liquid |
57184-23-3 | |
0.938 g/mL | |
87°C (189°F) | |
1.495 | |
5 g | |
LRPGNFROBDUREU-UHFFFAOYSA-N | |
1-(cyclohexylmethyl)piperazine | |
2735885 | |
97% |
Chemical Identifiers
57184-23-3 | |
182.31 | |
LRPGNFROBDUREU-UHFFFAOYSA-N | |
1-(cyclohexylmethyl)piperazine |
C11H22N2 | |
MFCD01075237 | |
2735885 | |
C(C1CCCCC1)N1CCNCC1 |
Safety and Handling
GHS H Statement
H318-H335-H227-H315
Causes serious eye damage.
May cause respiratory irritation.
Combustible liquid.
Causes skin irritation.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H318-H335
TSCA : No
Recommended Storage : Ambient temperatures