missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(Cyclohexylmethyl)piperazine, 97%

Manufacturer:  Alfa Aesar™ H55491MD

 View more versions of this product

Catalog No. AAH55491MD



Specifications

Specifications

1-(Cyclohexylmethyl)piperazine
87°C (189°F)
230°C to 231°C
250mg
C11H22N2
1-cyclohexyl-methyl piperazine
C1CCC(CC1)CN2CCNCC2
182.311
182.31
97%
0.938 g/mL
1.495
Colorless
57184-23-3
MFCD01075237
LRPGNFROBDUREU-UHFFFAOYSA-N
1-(cyclohexylmethyl)piperazine
2735885
Liquid

Chemical Identifiers

57184-23-3
182.311
LRPGNFROBDUREU-UHFFFAOYSA-N
2735885
C1CCC(CC1)CN2CCNCC2
C11H22N2
MFCD01075237
1-cyclohexyl-methyl piperazine
1-(cyclohexylmethyl)piperazine
Safety and Handling

Safety and Handling

GHS H Statement
H318-H335-H227-H315
Causes serious eye damage.
May cause respiratory irritation.
Combustible liquid.
Causes skin irritation.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H318-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents