Please login to your online account to display your discounted pricing

1-Cyclohexyl-4-oxo-1,4-dihydroquinoline-3-carboxylic acid, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 1-Cyclohexyl-4-oxo-1, 4-dihydroquinoline-3-carboxylic acid
CAS 135906-00-2
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C16H17NO3
Formula Weight 271.31
MDL Number MFCD00277770
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RJC01923EA

  SDS
fisher scientific company
RJC01923EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 271.31
Quantity 10g
Assay 97%
CAS Min % 97.0
CAS 135906-00-2
Assay Percent Range 97%
Chemical Name or Material 1-Cyclohexyl-4-oxo-1, 4-dihydroquinoline-3-carboxylic acid
MDL Number MFCD00277770
Packaging Amber Glass Bottle
Molecular Formula C16H17NO3