Please login to your online account to display your discounted pricing

1-Cyclohexyl-4-oxo-1,4-dihydroquinoline-3-carboxylic acid, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  RJC01923EA

 View more versions of this product

Catalog No. RJC01923EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 271.31
Quantity 10g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 1-Cyclohexyl-4-oxo-1, 4-dihydroquinoline-3-carboxylic acid
Assay Percent Range 97%
CAS 135906-00-2
Packaging Amber Glass Bottle
Molecular Formula C16H17NO3
MDL Number MFCD00277770

Specifications

Chemical Name or Material 1-Cyclohexyl-4-oxo-1, 4-dihydroquinoline-3-carboxylic acid
CAS 135906-00-2
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C16H17NO3
Formula Weight 271.31
MDL Number MFCD00277770