Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Cyclohexyl-2-buten-1-ol, tech. 90%

$53.49 - $53.49

Chemical Identifiers

Chemical Name or Material 1-Cyclohexyl-2-buten-1-ol
CAS 18736-82-8
Purity Grade Technical Grade
Molecular Formula C10H18O
Formula Weight 154.25
MDL Number MFCD00060819
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2050106

  SDS
alfa aesar™
B2050106
5g Each for $53.49
Description & Specifications

Specifications

Formula Weight 154.25
Chemical Name or Material 1-Cyclohexyl-2-buten-1-ol
Assay 90%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C10H18O
Purity Grade Technical Grade
Quantity 5g
MDL Number MFCD00060819
Health Hazard 1 Warning
CAS 18736-82-8
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.