Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Cyclohexyl-1-propanol, 97%

$65.73 - $65.73

Chemical Identifiers

Chemical Name or Material 1-Cyclohexyl-1-propanol
CAS 17264-02-7
Molecular Formula C9H18O
Formula Weight 142.24
MDL Number MFCD00001476
Synonym Cyclohexyl ethyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2049706

  SDS
alfa aesar™
B2049706
5g Each for $65.73
Description & Specifications

Specifications

Formula Weight 142.24
Chemical Name or Material 1-Cyclohexyl-1-propanol
TSCA TSCA
Assay 97%
Molecular Formula C9H18O
MDL Number MFCD00001476
CAS 17264-02-7
Synonym Cyclohexyl ethyl carbinol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 84°C (183°F)
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.