Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Cyclohexyl-1-pentanol, 98%

$50.78 - $192.86

Chemical Identifiers

Chemical Name or Material 1-Cyclohexyl-1-pentanol
CAS 7338-43-4
Molecular Formula C11H22O
Formula Weight 170.3
MDL Number MFCD00045483
Synonym n-Butyl cyclohexyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2019506

  SDS
alfa aesar™
B2019506
5g Each for $50.78

AAB2019514

  SDS
alfa aesar™
B2019514
25g Each for $192.86
Description & Specifications

Specifications

Formula Weight 170.3
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Synonym n-Butyl cyclohexyl carbinol
Assay 98%
Chemical Name or Material 1-Cyclohexyl-1-pentanol
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 7338-43-4
Quantity 5g
Molecular Formula C11H22O
MDL Number MFCD00045483
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.