Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Cyclohexyl-1-butanol, 98%

$50.78 - $192.86

Chemical Identifiers

Chemical Name or Material 1-Cyclohexyl-1-butanol
CAS 4352-42-5
Molecular Formula C10H20O
Formula Weight 156.27
MDL Number MFCD00019360
Synonym n-Propyl cyclohexyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2009006

  SDS
alfa aesar™
B2009006
5g Each for $50.78

AAB2009014

  SDS
alfa aesar™
B2009014
25g Each for $192.86
Description & Specifications

Specifications

Formula Weight 156.27
MDL Number MFCD00019360
Chemical Name or Material 1-Cyclohexyl-1-butanol
Health Hazard 1 Warning
Assay 98%
Synonym n-Propyl cyclohexyl carbinol
Quantity 5g
Molecular Formula C10H20O
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 103° to 104°C (4mmHg)
Flash Point 108°C (228°F)
CAS 4352-42-5
TSCA TSCA
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.