missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Cyclohexenylacetonitrile 92%, ACROS Organics™

Chemical Identifiers

CAS 6975-71-9
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
InChI Key OYEXEQFKIPJKJK-UHFFFAOYSA-N
Synonym 1-cyclohexene-1-acetonitrile, 2-cyclohex-1-en-1-yl acetonitrile, 1-cyclohexenylacetonitrile, cyclohexenylacetonitrile, cyclohex-1-enyl-acetonitrile, 1-cyclohexen-1-ylacetonitrile, 1-cyclohexenacetonitrile, 1-cyclohexeneacetonitrile, 2-cyclohexen-1-yl acetonitrile, 2-cyclohex-1-enylethanenitrile
PubChem CID 81461
IUPAC Name 2-(cyclohexen-1-yl)acetonitrile
SMILES C1CCC(=CC1)CC#N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC111250100 SDS View Documents Acros Organics
AC111250100
10g Glass bottle N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

6975-71-9
121.183
1-cyclohexene-1-acetonitrile, 2-cyclohex-1-en-1-yl acetonitrile, 1-cyclohexenylacetonitrile, cyclohexenylacetonitrile, cyclohex-1-enyl-acetonitrile, 1-cyclohexen-1-ylacetonitrile, 1-cyclohexenacetonitrile, 1-cyclohexeneacetonitrile, 2-cyclohexen-1-yl acetonitrile, 2-cyclohex-1-enylethanenitrile
2-(cyclohexen-1-yl)acetonitrile
C8H11N
OYEXEQFKIPJKJK-UHFFFAOYSA-N
81461
C1CCC(=CC1)CC#N

Specifications

Authentic
Yellow to Orange
6975-71-9
100.0
C8H11N
1-cyclohexene-1-acetonitrile, 2-cyclohex-1-en-1-yl acetonitrile, 1-cyclohexenylacetonitrile, cyclohexenylacetonitrile, cyclohex-1-enyl-acetonitrile, 1-cyclohexen-1-ylacetonitrile, 1-cyclohexenacetonitrile, 1-cyclohexeneacetonitrile, 2-cyclohexen-1-yl acetonitrile, 2-cyclohex-1-enylethanenitrile
C1CCC(=CC1)CC#N
121.183
121.18
≥88.0%
Glass bottle
10g
88.0
90%
C6H9CH2CN
OYEXEQFKIPJKJK-UHFFFAOYSA-N
2-(cyclohexen-1-yl)acetonitrile
81461
Liquid
1-Cyclohexenylacetonitrile, 92%
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 230-235-6

TSCA : TSCA