missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Cyclohexenylacetonitrile 92%, ACROS Organics™

Manufacturer:  Acros Organics AC111250100

 View more versions of this product

Catalog No. AC111250100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1-Cyclohexenylacetonitrile
Glass bottle
10g
6975-71-9
100.0
C8H11N
1-cyclohexene-1-acetonitrile, 2-cyclohex-1-en-1-yl acetonitrile, 1-cyclohexenylacetonitrile, cyclohexenylacetonitrile, cyclohex-1-enyl-acetonitrile, 1-cyclohexen-1-ylacetonitrile, 1-cyclohexenacetonitrile, 1-cyclohexeneacetonitrile, 2-cyclohexen-1-yl acetonitrile, 2-cyclohex-1-enylethanenitrile
C1CCC(=CC1)CC#N
121.183
121.18
≥88.0%
Authentic
Yellow to Orange
92%
88.0
90%
C6H9CH2CN
OYEXEQFKIPJKJK-UHFFFAOYSA-N
2-(cyclohexen-1-yl)acetonitrile
81461
Liquid

Chemical Identifiers

6975-71-9
121.183
1-cyclohexene-1-acetonitrile, 2-cyclohex-1-en-1-yl acetonitrile, 1-cyclohexenylacetonitrile, cyclohexenylacetonitrile, cyclohex-1-enyl-acetonitrile, 1-cyclohexen-1-ylacetonitrile, 1-cyclohexenacetonitrile, 1-cyclohexeneacetonitrile, 2-cyclohexen-1-yl acetonitrile, 2-cyclohex-1-enylethanenitrile
2-(cyclohexen-1-yl)acetonitrile
C8H11N
OYEXEQFKIPJKJK-UHFFFAOYSA-N
81461
C1CCC(=CC1)CC#N
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 230-235-6

TSCA : TSCA

SDS Documents