Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Cyclohexenyl trifluoromethanesulfonate, 97%

$52.74 - $52.74

Chemical Identifiers

Chemical Name or Material 1-Cyclohexenyl trifluoromethanesulfonate
CAS 28075-50-5
Formula Weight 230.2
MDL Number MFCD06411281
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5587903

  SDS
alfa aesar™
H5587903
1g Each for $52.74

AAH5587906

  SDS
alfa aesar™
H5587906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5587914

  SDS
alfa aesar™
H5587914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 84° to 87°C (4mmHg)
Formula Weight 230.2
MDL Number MFCD06411281
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Health Hazard 1 Warning
Color Colorless to yellow
Chemical Name or Material 1-Cyclohexenyl trifluoromethanesulfonate
Assay 97%
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Physical Form Liquid
Quantity 1g
Density 1.315g/mL
CAS 28075-50-5

Applications
Used as a pharmaceutical intermediate.

Notes
Store at 4°C.

Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.