Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Cyclohexenyl acetate, 96%

$343.26 - $343.26

Chemical Identifiers

Chemical Name or Material 1-Cyclohexenyl acetate
CAS 1424-22-2
Molecular Formula C8H12O2
Formula Weight 140.18
MDL Number MFCD00013774
Beilstein 878815
Synonym 1-Acetoxycyclohexene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0497018

  SDS
alfa aesar™
L0497018
50g Each for $343.26
Description & Specifications

Specifications

Formula Weight 140.18
Chemical Name or Material 1-Cyclohexenyl acetate
Molecular Formula C8H12O2
MDL Number MFCD00013774
CAS 1424-22-2
Synonym 1-Acetoxycyclohexene
Health Hazard 1 Warning
Assay 96%
Beilstein 878815
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Boiling Point 76° to 77°C (17mmHg)
Quantity 50g
Flash Point 65°C (149°F)
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.