Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Cyclohexene-1-boronic acid pinacol ester, 95%

$112.39 - $328.47

Chemical Identifiers

Chemical Name or Material 1-Cyclohexene-1-boronic acid pinacol ester
CAS 141091-37-4
Molecular Formula C12H21BO2
Formula Weight 208.1
MDL Number MFCD05663845
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5103603

  SDS
alfa aesar™
H5103603
1g Each for $328.47

AAH51036MD

  SDS
alfa aesar™
H51036MD
250mg Each for $112.39
Description & Specifications

Specifications

Formula Weight 208.1
Flash Point 103°C (217°F)
Molecular Formula C12H21BO2
MDL Number MFCD05663845
CAS 141091-37-4
Assay 95%
Chemical Name or Material 1-Cyclohexene-1-boronic acid pinacol ester
Recommended Storage 2° to 8°C
Quantity 1g
Safety and Handling

2° to 8°C