missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Cyclohexene-1-acetic acid

Manufacturer:  Alfa Aesar™ 1946203

 View more versions of this product

Catalog No. AA1946203



Specifications

Specifications

1-Cyclohexene-1-acetic acid
1.4775
33°C
18294-87-6
MFCD00015482
KDFBPHXESBPHTK-UHFFFAOYSA-N
2-(cyclohexen-1-yl)acetic acid
86699
Liquid/Low Melting Solid
1.04g/mL
140°C to 142°C (25mmHg)
1g
C8H12O2
1-cyclohexenylacetic acid, 1-cyclohexene-1-acetic acid, 2-cyclohex-1-en-1-yl acetic acid, cyclohex-1-enylacetic acid, 1-cyclohexene-1-aceticacid, 2-cyclohexen-1-yl acetic acid, 2-cyclohex-1-enylacetic acid, cyclohex-1-en-1-ylacetic acid, 1-cyclohexenylacetate, cyclohexenylacetic acid
C1CCC(=CC1)CC(=O)O
140.182
140.18

Chemical Identifiers

18294-87-6
140.182
KDFBPHXESBPHTK-UHFFFAOYSA-N
86699
C1CCC(=CC1)CC(=O)O
C8H12O2
MFCD00015482
1-cyclohexenylacetic acid, 1-cyclohexene-1-acetic acid, 2-cyclohex-1-en-1-yl acetic acid, cyclohex-1-enylacetic acid, 1-cyclohexene-1-aceticacid, 2-cyclohexen-1-yl acetic acid, 2-cyclohex-1-enylacetic acid, cyclohex-1-en-1-ylacetic acid, 1-cyclohexenylacetate, cyclohexenylacetic acid
2-(cyclohexen-1-yl)acetic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents