missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Cyanomethyl-4,5-dicyano-2-(4-pyridyl)imidazole, 97%

Manufacturer:  Alfa Aesar™ H5425406

 View more versions of this product

Catalog No. AAH5425406

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1-Cyanomethyl-4,5-dicyano-2-(4-pyridyl)imidazole
1347815-36-4
MFCD20265215
1-cyanomethyl-2-pyridin-4-yl imidazole-4,5-dicarbonitrile
C1=CN=CC=C1C2=NC(=C(N2CC#N)C#N)C#N
234.222
234.22
5g
C12H6N6
UN3439
QPUUMBCRXZGNEF-UHFFFAOYSA-N
1-(cyanomethyl)-2-pyridin-4-ylimidazole-4,5-dicarbonitrile
73995714
97%

Chemical Identifiers

1347815-36-4
234.222
QPUUMBCRXZGNEF-UHFFFAOYSA-N
73995714
C1=CN=CC=C1C2=NC(=C(N2CC#N)C#N)C#N
C12H6N6
MFCD20265215
1-cyanomethyl-2-pyridin-4-yl imidazole-4,5-dicarbonitrile
1-(cyanomethyl)-2-pyridin-4-ylimidazole-4,5-dicarbonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Documents