missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Cyanomethyl-2-(4-cyanophenyl)-4,5-dicyanoimidazole, 97%

Manufacturer:  Alfa Aesar™ H5446903

 View more versions of this product

Catalog No. AAH5446903

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1-Cyanomethyl-2-(4-cyanophenyl)-4,5-dicyanoimidazole
1347815-43-3
MFCD20265274
1-cyanomethyl-2-4-cyanophenyl imidazole-4,5-dicarbonitrile
C1=CC(=CC=C1C#N)C2=NC(=C(N2CC#N)C#N)C#N
258.244
258.24
1g
C14H6N6
UN3439
OAMIMEALBFOHIE-UHFFFAOYSA-N
1-(cyanomethyl)-2-(4-cyanophenyl)imidazole-4,5-dicarbonitrile
73995824
97%

Chemical Identifiers

1347815-43-3
258.244
OAMIMEALBFOHIE-UHFFFAOYSA-N
73995824
C1=CC(=CC=C1C#N)C2=NC(=C(N2CC#N)C#N)C#N
C14H6N6
MFCD20265274
1-cyanomethyl-2-4-cyanophenyl imidazole-4,5-dicarbonitrile
1-(cyanomethyl)-2-(4-cyanophenyl)imidazole-4,5-dicarbonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P361-P363-P501c

H301+H311-H315-H319-H330-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents