missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Cyanomethyl-2-(3-cyanophenyl)-4,5-dicyanoimidazole, 97%

Manufacturer:  Alfa Aesar™ H54024MD

 View more versions of this product

Catalog No. AAH54024MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1-Cyanomethyl-2-(3-cyanophenyl)-4,5-dicyanoimidazole
1347815-42-2
MFCD20265160
1-cyanomethyl-2-3-cyanophenyl imidazole-4,5-dicarbonitrile
C1=CC(=CC(=C1)C2=NC(=C(N2CC#N)C#N)C#N)C#N
258.244
258.24
250mg
C14H6N6
UN3439
KQHKTLGFDJPVEP-UHFFFAOYSA-N
1-(cyanomethyl)-2-(3-cyanophenyl)imidazole-4,5-dicarbonitrile
73995593
97%

Chemical Identifiers

1347815-42-2
258.244
KQHKTLGFDJPVEP-UHFFFAOYSA-N
73995593
C1=CC(=CC(=C1)C2=NC(=C(N2CC#N)C#N)C#N)C#N
C14H6N6
MFCD20265160
1-cyanomethyl-2-3-cyanophenyl imidazole-4,5-dicarbonitrile
1-(cyanomethyl)-2-(3-cyanophenyl)imidazole-4,5-dicarbonitrile
Safety and Handling

Safety and Handling

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents