Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(Cyanoacetyl)piperidine, 98%

$46.80 - $46.80

Chemical Identifiers

Chemical Name or Material 1-(Cyanoacetyl)piperidine
CAS 15029-30-8
Molecular Formula C8H12N2O
Formula Weight 152.2
MDL Number MFCD00006487
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2053206

 
alfa aesar™
B2053206
5g Each for $46.80
Description & Specifications

Specifications

Formula Weight 152.2
Chemical Name or Material 1-(Cyanoacetyl)piperidine
Odor Odorless
Assay 98%
Molecular Formula C8H12N2O
MDL Number MFCD00006487
CAS 15029-30-8
Health Hazard 1 Warning
Melting Point 86° to 89°C
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.