Please login to your online account to display your discounted pricing

1-Cyano-4-dimethylaminopyridinium tetrafluoroborate, +97%, ACROS Organics™


Quantity 100mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC397421000

 View more versions of this product

Catalog No. AC397421000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Assay ≥97.0%
Formula Weight 234.99
Quantity 100mg
Color White to yellow
CAS Min % 97.0
CAS Max % 100.0
Infrared Spectrum Authentic
Chemical Name or Material 1-Cyano-4-dimethylaminopyridinium tetrafluoroborate
Molecular Formula C8H10BF4N3
CAS 59016-56-7
Assay Percent Range 97+%
Health Hazard 1 GHS Signal Word: Warning
MDL Number MFCD00011998
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 1-Cyano-4-dimethylaminopyridinium tetrafluoroborate
CAS 59016-56-7
CAS Min % 97.0
CAS Max % 100.0
Assay ≥97.0%
Assay Percent Range 97+%
Molecular Formula C8H10BF4N3
Formula Weight 234.99
MDL Number MFCD00011998
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.

SDS & Safety Documents