missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Chlorotetradecane, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 294622500
Specifications
1-Chlorotetradecane | |
Glass bottle | |
Colorless | |
98% | |
97.5 | |
98% | |
CH3(CH2)13Cl | |
1-chloranyltetradecane, 1-chloro-tetradecane, acmc-1cmeh, chlorotetradecane, myristyl chloride, n-tetradecyl chloride, tetradecane, 1-chloro, tetradecane, chloro, tetradecyl chloride, tetradecylchloride | |
CCCCCCCCCCCCCCCl | |
232.836 | |
232.84 | |
≥97.5% |
Authentic | |
1.4460 to 1.4480 (20°C, 589nm) | |
250mL | |
2425-54-9 | |
100.0 | |
C14H29Cl | |
MFCD00000958 | |
RNHWYOLIEJIAMV-UHFFFAOYSA-N | |
1-chlorotetradecane | |
17043 | |
Liquid |
Chemical Identifiers
2425-54-9 | |
232.836 | |
RNHWYOLIEJIAMV-UHFFFAOYSA-N | |
17043 | |
CCCCCCCCCCCCCCCl |
C14H29Cl | |
MFCD00000958 | |
1-chloranyltetradecane, 1-chloro-tetradecane, acmc-1cmeh, chlorotetradecane, myristyl chloride, n-tetradecyl chloride, tetradecane, 1-chloro, tetradecane, chloro, tetradecyl chloride, tetradecylchloride | |
1-chlorotetradecane |
Safety and Handling
EINECSNumber : 219-368-0
TSCA : TSCA