missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Chlorotetradecane, 98%, ACROS Organics™

$71.92 - $71.92

Chemical Identifiers

CAS 2425-54-9
Molecular Formula C14H29Cl
Molecular Weight (g/mol) 232.836
MDL Number MFCD00000958
InChI Key RNHWYOLIEJIAMV-UHFFFAOYSA-N
Synonym myristyl chloride, tetradecane, 1-chloro, tetradecyl chloride, n-tetradecyl chloride, tetradecane, chloro, chlorotetradecane, 1-chloranyltetradecane, tetradecylchloride, 1-chloro-tetradecane, acmc-1cmeh
PubChem CID 17043
IUPAC Name 1-chlorotetradecane
SMILES CCCCCCCCCCCCCCCl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC294622500 SDS View Documents Acros Organics
294622500
250mL Each for $71.92
Specifications

Chemical Identifiers

2425-54-9
232.836
RNHWYOLIEJIAMV-UHFFFAOYSA-N
17043
CCCCCCCCCCCCCCCl
C14H29Cl
MFCD00000958
myristyl chloride, tetradecane, 1-chloro, tetradecyl chloride, n-tetradecyl chloride, tetradecane, chloro, chlorotetradecane, 1-chloranyltetradecane, tetradecylchloride, 1-chloro-tetradecane, acmc-1cmeh
1-chlorotetradecane

Specifications

Authentic
1.4460 to 1.4480 (20°C, 589nm)
250mL
97.5
98%
CH3(CH2)13Cl
myristyl chloride, tetradecane, 1-chloro, tetradecyl chloride, n-tetradecyl chloride, tetradecane, chloro, chlorotetradecane, 1-chloranyltetradecane, tetradecylchloride, 1-chloro-tetradecane, acmc-1cmeh
CCCCCCCCCCCCCCCl
232.836
232.84
≥97.5%
Glass bottle
Colorless
2425-54-9
100.0
C14H29Cl
MFCD00000958
RNHWYOLIEJIAMV-UHFFFAOYSA-N
1-chlorotetradecane
17043
Liquid
1-Chlorotetradecane, 98%
Safety and Handling

Safety and Handling

EINECSNumber : 219-368-0

TSCA : TSCA