Please login to your online account to display your discounted pricing

1-Chloropropane, 99%, ACROS Organics™


Quantity 500mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC109955000

 View more versions of this product

Catalog No. AC109955000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥98.5%
Formula Weight 78.54
Quantity 500mL
Color Clear colorless
Density 0.892g/mL
Beilstein 01, 104
Merck Index 15, 7960
CAS Max % 100.0
Molecular Formula C3H7Cl
Linear Formula CH3CH2CH2Cl
CAS 540-54-5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 1-chloropropane
Packaging Glass bottle
Refractive Index 1.3870 to 1.3890 (20°C, 589nm)
MDL Number MFCD00000995
Synonym n-Propyl chloride
CAS Min % 98.5
Infrared Spectrum Authentic
Assay Percent Range 99%
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Highly flammable liquid and vapor.

Specifications

Chemical Name or Material 1-chloropropane
CAS 540-54-5
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C3H7Cl
Linear Formula CH3CH2CH2Cl
Formula Weight 78.54
MDL Number MFCD00000995
Synonym n-Propyl chloride
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Highly flammable liquid and vapor.

SDS & Safety Documents