missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Chloropinacolone, 95%

$20.30 - $20.30

Chemical Identifiers

CAS 13547-70-1
Molecular Formula C6H11ClO
Linear Formula (CH3)3CCOCH2Cl
Formula Weight 134.59
MDL Number MFCD00035688
UN Number UN3382
Synonym 1-Chloro-3,3-dimethyl-2-butanone
Chemical Name or Material 1-Chloropinacolone
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1657614 SDS Alfa Aesar™
A1657614
25g Each for $20.30
AAA1657622 Alfa Aesar™
A1657622
100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAA1657636 Alfa Aesar™
A1657636
500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
1-Chloropinacolone is used as an intermediate for triazole compounds ( a five-membered ring structure compound contains three nitrogens and two carbons in the ring) which have biological activities such as antiviral, antibacterial, antifungal and antituberculous.

Solubility
Not miscible in water.

Notes
Incompatible with oxidizing agents and bases. Store in a cool, dry, conditions in well sealed container. Store at 4°C.
Description & Specifications

Chemical Identifiers

13547-70-1
(CH3)3CCOCH2Cl
MFCD00035688
1-Chloro-3,3-dimethyl-2-butanone
C6H11ClO
134.59
UN3382
1-Chloropinacolone

Specifications

67°C (153°F)
170° to 173°C
25g
C6H11ClO
134.59
UN3382
1-Chloropinacolone
Pungent
−13°C
13547-70-1
(CH3)3CCOCH2Cl
MFCD00035688
1-Chloro-3,3-dimethyl-2-butanone
95%

Safety and Handling

GHS H Statement
H301-H311-H330-H227-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280-P305+P351+P338-P304+P340-P309-P310-P302+P352
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, N.O.S.

RTECSNumber : EL7078000

TSCA : TSCA

Recommended Storage : 2° to 8°C