Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Chloropentane, 98%

Chemical Identifiers

Chemical Name or Material 1-Chloropentane
CAS 543-59-9
Molecular Formula C5H11Cl
Linear Formula CH3(CH2)4Cl
Formula Weight 106.6
MDL Number MFCD00001015
UN Number UN1107
Merck Index 14,606
Synonym n-Pentyl chloride; n-Amyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB247330B

  SDS
alfa aesar™
B247330B
1000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2473318

  SDS
alfa aesar™
B2473318
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2473330

  SDS
alfa aesar™
B2473330
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 106.6
Chemical Name or Material 1-Chloropentane
TSCA TSCA
Assay 98%
Molecular Formula C5H11Cl
Linear Formula CH3(CH2)4Cl
MDL Number MFCD00001015
CAS 543-59-9
UN Number UN1107
Merck Index 14,606
Synonym n-Pentyl chloride; n-Amyl chloride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H302-H312-H332
Highly flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P273-P403
Avoid release to the environment.
Store in a well-ventilated place.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMYL CHLORIDES
Flash Point 3°C (37°F)
Odor Sweet
Boiling Point 106° to 108°C
Melting Point −99°C
Quantity 1000g
Safety and Handling

Danger

GHS P Statement
P273-P403
Avoid release to the environment.
Store in a well-ventilated place.

GHS H Statement
H225-H302-H312-H332
Highly flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.