Please login to your online account to display your discounted pricing

1-Chlorooctane 99%, ACROS Organics™


Quantity 500mL
Packaging Glass bottle

Manufacturer: fisher scientific acros  154715000

 View more versions of this product

Catalog No. AC154715000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4288 to 1.4308 (20°C, 589nm)
Color Clear
Quantity 500mL
Chemical Name or Material 1-Chlorooctane
Name Note 99%
CAS 111-85-3
CAS Min % 98.5
CAS Max % 100.0
Physical Form Clear liquid
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C8H17Cl
Linear Formula CH3(CH2)7Cl
Formula Weight 148.68g/mol
MDL Number MFCD00001022
Beilstein 01, 159
Synonym 1-Octylchloride
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECS Number 203-915-5
TSCA TSCA

Specifications

Chemical Name or Material 1-Chlorooctane
Name Note 99%
CAS 111-85-3
CAS Min % 98.5
CAS Max % 100.0
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C8H17Cl
Linear Formula CH3(CH2)7Cl
Formula Weight 148.68g/mol
MDL Number MFCD00001022
Synonym 1-Octylchloride
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Very toxic to aquatic life with long lasting effects.

SDS & Safety Documents