missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Chlorooctane 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 154711000

 View more versions of this product

Catalog No. AC154711000


 

Specifications Description & Specifications

Specifications

≥98.5%
Liquid
Clear
100.0
99%
1-Chlorooctane
111-85-3
99%
Glass bottle
Authentic
148.68
100mL
01, 159
1.4288 to 1.4308 (20°C, 589nm)
CH3(CH2)7Cl
98.5
MFCD00001022
C8H17Cl
1-Octylchloride

Chemical Identifiers

111-85-3
100.0
C8H17Cl
148.68
1-Octylchloride
≥98.5%
98.5
99%
CH3(CH2)7Cl
MFCD00001022
1-Chlorooctane, 99%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-915-5

TSCA : TSCA