Please login to your online account to display your discounted pricing

1-Chlorooctane 99%, ACROS Organics™


Quantity 5mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  154710050

 View more versions of this product

Catalog No. AC154710050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 148.68
Physical Form Liquid
Quantity 5mL
Color Clear
Beilstein 01, 159
EINECS Number 203-915-5
TSCA TSCA
CAS Max % 100.0
Chemical Name or Material 1-Chlorooctane
Health Hazard 3 GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C8H17Cl
CAS 111-85-3
MDL Number MFCD00001022
Assay Percent Range 99%
Linear Formula CH3(CH2)7Cl
CAS Min % 98.5
Name Note 99%
Refractive Index 1.4288 to 1.4308 (20°C, 589nm)
Packaging Glass bottle
Infrared Spectrum Authentic
Synonym 1-Octylchloride
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Very toxic to aquatic life with long lasting effects.

Specifications

Chemical Name or Material 1-Chlorooctane
Name Note 99%
CAS 111-85-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H17Cl
Linear Formula CH3(CH2)7Cl
Formula Weight 148.68
MDL Number MFCD00001022
Synonym 1-Octylchloride
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Very toxic to aquatic life with long lasting effects.

SDS & Safety Documents