Please login to your online account to display your discounted pricing

1-Chlorooctane 99%, ACROS Organics™

$22.28 - $22.28

Chemical Identifiers

Chemical Name or Material 1-Chlorooctane
Name Note 99%
CAS 111-85-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H17Cl
Linear Formula CH3(CH2)7Cl
Formula Weight 148.68
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC154710050

  SDS
thermo fisher scientific
154710050
Glass bottle 5mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC154711000

  SDS
thermo fisher scientific
154711000
Glass bottle 100mL Each for $22.28
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 148.68
Physical Form Liquid
Quantity 5mL
Color Clear
Beilstein 01, 159
EINECS Number 203-915-5
TSCA TSCA
CAS Max % 100.0
Chemical Name or Material 1-Chlorooctane
Health Hazard 3 GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C8H17Cl
CAS 111-85-3
MDL Number MFCD00001022
Assay Percent Range 99%
Linear Formula CH3(CH2)7Cl
CAS Min % 98.5
Name Note 99%
Refractive Index 1.4288 to 1.4308 (20°C, 589nm)
Packaging Glass bottle
Infrared Spectrum Authentic
Synonym 1-Octylchloride
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Very toxic to aquatic life with long lasting effects.