missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorooctadecane, 94%

CAS: 3386-33-2 | C18H37Cl | 288.94 g/mol

$70.78 - $605.49

Chemical Identifiers

CAS 3386-33-2
Molecular Formula C18H37Cl
Molecular Weight (g/mol) 288.94
MDL Number MFCD00000960
InChI Key VUQPJRPDRDVQMN-UHFFFAOYSA-N
Synonym octadecyl chloride, octadecane, 1-chloro, n-octadecyl chloride, 1-chloroctadecane, chlorooctadecane, octadecane, chloro, unii-73h8vd533i, 1-chloranyloctadecane, stearylchloride, acmc-1cnf6
PubChem CID 18815
IUPAC Name 1-chlorooctadecane
SMILES CCCCCCCCCCCCCCCCCCCl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2490014
View Documents
Thermo Scientific Chemicals
B2490014
25 g
Each for $70.78
Only null left
 
AAB2490022
View Documents
Thermo Scientific Chemicals
B2490022
100 g
Each for $165.95
Only null left
 
AAB2490036
View Documents
Thermo Scientific Chemicals
B2490036
500 g
Each for $605.49
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3386-33-2
288.94
VUQPJRPDRDVQMN-UHFFFAOYSA-N
18815
CCCCCCCCCCCCCCCCCCCl
C18H37Cl
MFCD00000960
octadecyl chloride, octadecane, 1-chloro, n-octadecyl chloride, 1-chloroctadecane, chlorooctadecane, octadecane, chloro, unii-73h8vd533i, 1-chloranyloctadecane, stearylchloride, acmc-1cnf6
1-chlorooctadecane

Specifications

3386-33-2
0.849
>110°C (230°F)
1.451
MFCD00000960
Moisture sensitive
VUQPJRPDRDVQMN-UHFFFAOYSA-N
1-chlorooctadecane
18815
94%
20°C to 23°C
157°C to 158°C (1.5 mmHg)
C18H37Cl
CH3(CH2)17Cl
25 g
octadecyl chloride, octadecane, 1-chloro, n-octadecyl chloride, 1-chloroctadecane, chlorooctadecane, octadecane, chloro, unii-73h8vd533i, 1-chloranyloctadecane, stearylchloride, acmc-1cnf6
CCCCCCCCCCCCCCCCCCCl
288.94
288.95
1-Chlorooctadecane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 222-207-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only