Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Chlorononane, 98%

$47.41 - $47.41

Chemical Identifiers

Chemical Name or Material 1-Chlorononane
CAS 2473-01-0
Molecular Formula C9H19Cl
Linear Formula CH3(CH2)8Cl
Formula Weight 162.7
MDL Number MFCD00001023
Beilstein 1733625
Synonym n-Nonyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0990909

  SDS
alfa aesar™
L0990909
10g Each for $47.41
Description & Specifications

Specifications

Formula Weight 162.7
Chemical Name or Material 1-Chlorononane
TSCA TSCA
Assay 98%
Molecular Formula C9H19Cl
Linear Formula CH3(CH2)8Cl
MDL Number MFCD00001023
CAS 2473-01-0
Beilstein 1733625
Synonym n-Nonyl chloride
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 74°C (165°F)
Boiling Point 201° to 202°C
Quantity 10g
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.