missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Chlorononane, 98%

Manufacturer:  Alfa Aesar™ L0990918

 View more versions of this product

Catalog No. AAL0990918



Specifications

Specifications

1-Chlorononane
201°C to 202°C
2473-01-0
CH3(CH2)8Cl
1733625
RKAMCQVGHFRILV-UHFFFAOYSA-N
1-chlorononane
17185
98%
74°C (165°F)
50g
C9H19Cl
MFCD00001023
nonyl chloride, nonane, 1-chloro, n-nonyl chloride, nonane,1-chloro, tetrapropenyl chloride, 1-propene, tetramer, chlorinated, acmc-209gec, 1-chlorononane, dsstox_cid_29111, dsstox_rid_83330
CCCCCCCCCCl
162.701
162.7

Chemical Identifiers

2473-01-0
162.701
RKAMCQVGHFRILV-UHFFFAOYSA-N
17185
CCCCCCCCCCl
C9H19Cl
MFCD00001023
nonyl chloride, nonane, 1-chloro, n-nonyl chloride, nonane,1-chloro, tetrapropenyl chloride, 1-propene, tetramer, chlorinated, acmc-209gec, 1-chlorononane, dsstox_cid_29111, dsstox_rid_83330
1-chlorononane
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents