missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Chloronaphthalene, tech. 85%, remainder 2-chloronaphthalene

Manufacturer:  Alfa Aesar™ A1502930

 View more versions of this product

Catalog No. AAA1502930



Description

Description

Applications
1-Chloronaphthalene was used as solvent additive in the fabrication of fullerene based photovoltaic devices. It was used in preparation of polyvinyl chloride based membranes of 5,10,15,20-tetrakis(4-methoxyphenyl)porphyrinatocobalt(II) for arsenite determination. Also used in the synthesis of pharmaceutical compounds such as aryl chlorides.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

1-Chloronaphthalene
259°C
250g
remainder 2-chloronaphthalene
MFCD00003874
14,2149
JTPNRXUCIXHOKM-UHFFFAOYSA-N
1-chloronaphthalene
7003
85%
121°C (249°F)
~-8°C
90-13-1
C10H7Cl
970836
alpha-chloronaphthalene, naphthalene, 1-chloro, 1-naphthyl chloride, chloronaphthalene, 1-chloro-naphthalene, alpha-naphthyl chloride, 1-chlornaftalen, 1-chloro naphthalene, alpha-chlornaphthalene, .alpha.-chloronaphthalene
C1=CC=C2C(=C1)C=CC=C2Cl
162.616
162.62
Technical

Chemical Identifiers

90-13-1
162.616
JTPNRXUCIXHOKM-UHFFFAOYSA-N
7003
C1=CC=C2C(=C1)C=CC=C2Cl
C10H7Cl
MFCD00003874
alpha-chloronaphthalene, naphthalene, 1-chloro, 1-naphthyl chloride, chloronaphthalene, 1-chloro-naphthalene, alpha-naphthyl chloride, 1-chlornaftalen, 1-chloro naphthalene, alpha-chlornaphthalene, .alpha.-chloronaphthalene
1-chloronaphthalene
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : QJ2100000

TSCA : TSCA

Documents