Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(Chloromethyl)naphthalene, 94%

Chemical Identifiers

Chemical Name or Material 1-(Chloromethyl)naphthalene
CAS 86-52-2
Molecular Formula C11H9Cl
Formula Weight 176.65g/mol
MDL Number MFCD00004042
UN Number UN3261
Beilstein 636885
Synonym 1-Naphthylmethyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2438522

 
alfa aesar™
B24385-22
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2438530

 
alfa aesar™
B24385-30
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2438536

 
alfa aesar™
B24385-36
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Odor Pungent
Melting Point 31° to 33°C
Boiling Point 291° to 292°C
Quantity 100g
Flash Point 132°C (270°F)
Chemical Name or Material 1-(Chloromethyl)naphthalene
CAS 86-52-2
Assay 94%
Molecular Formula C11H9Cl
Formula Weight 176.65g/mol
MDL Number MFCD00004042
UN Number UN3261
Beilstein 636885
Synonym 1-Naphthylmethyl chloride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
RTECS Number QJ2800000
TSCA TSCA

Applications
1-(Chloromethyl)naphthalene is a key material for synthesis dye pigment and as fluorescent brightening agent. It is employed in synthetic resin and medicine. Palladium-catalyzed nucleophilic dearomatization of chloromethyl naphthalene derivatives produce ortho- or para-substituted carbocycles in satisfactory to excellent yields.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents and bases.

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.