Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Chlorohexane, 96%

Chemical Identifiers

Chemical Name or Material 1-Chlorohexane
CAS 544-10-5
Molecular Formula C6H13Cl
Linear Formula CH3(CH2)5Cl
Formula Weight 120.62
MDL Number MFCD00001018
UN Number UN1993
Beilstein 1731289
Merck Index 14,2144
Synonym n-Hexyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2237918

  SDS
alfa aesar™
B2237918
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2237930

  SDS
alfa aesar™
B2237930
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 120.62
Chemical Name or Material 1-Chlorohexane
TSCA TSCA
Assay 96%
Molecular Formula C6H13Cl
Linear Formula CH3(CH2)5Cl
MDL Number MFCD00001018
CAS 544-10-5
UN Number UN1993
Beilstein 1731289
Merck Index 14,2144
Synonym n-Hexyl chloride
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Flash Point 26°C (78°F)
Boiling Point 132° to 134°C
Melting Point −94°C
Quantity 50g
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapour.