Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Chloroethyl chloroformate, 98%


Quantity 25g

Manufacturer: alfa aesar™  L1419714

 View more versions of this product

Catalog No. AAL1419714

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 142.97
Chemical Name or Material 1-Chloroethyl chloroformate
Molecular Formula C3H4Cl2O2
Synonym Chloroformic acid 1-chloroethyl ester
Assay 98%
MDL Number MFCD00000647
CAS 50893-53-3
UN Number UN2742
Beilstein 1747601
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H330-H314-H318-H226-H302
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROFORMATES, TOXIC, CORROSIVE, FLAMMABLE, N.O.S.
Flash Point 45°C (113°F)
Boiling Point 119° to 121°C
Quantity 25g
Melting Point −65°C

Specifications

Chemical Name or Material 1-Chloroethyl chloroformate
CAS 50893-53-3
Molecular Formula C3H4Cl2O2
Formula Weight 142.97
MDL Number MFCD00000647
UN Number UN2742
Beilstein 1747601
Synonym Chloroformic acid 1-chloroethyl ester

Applications
1-Chloroethyl chloroformate acts as a reagent in the selective N-dealkylation of tertiary amines. It is also used as a reactant in the preparation of alkyl 1-chloroethyl carbonates. Further, it is used for N-demethylation reaction in the determination of multiple drugs of abuse in biological fluids using capillary electrophoresis with native fluorescence and laser-induced fluorescence detection. It plays an important role as a catalyst for the chemoselective desilylation of silyl-protected alcohols. In addition to this, it is used to cleave benzhydryl groups from amines.

Solubility
Miscible with most organic solvents.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agent, strong base, alcohols and amines.

Accessories

Danger

GHS P Statement
P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H330-H314-H318-H226-H302
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.

2° to 8°C