Please login to your online account to display your discounted pricing

1-Chloroethyl chloroformate, 97+%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC314460250

 View more versions of this product

Catalog No. AC314460250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.0%
Physical Form Liquid
Formula Weight 142.97
Quantity 25g
Color Clear colorless
CAS Min % 97.0
CAS Max % 100.0
Chemical Name or Material 1-Chloroethyl chloroformate
Refractive Index 1.4220 to 1.4240 (20°C, 589nm)
Assay Percent Range 97+%
MDL Number MFCD00000647
CAS 50893-53-3
Health Hazard 1 GHS Signal Word: Danger
Packaging Glass bottle
Molecular Formula C3H4Cl2O2
Linear Formula ClCOCH(Cl)CH3
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Fatal if inhaled.
Flammable liquid and vapor.
Contact with water liberates toxic gas.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 1-Chloroethyl chloroformate
CAS 50893-53-3
CAS Min % 97.0
CAS Max % 100.0
Assay ≥97.0%
Assay Percent Range 97+%
Molecular Formula C3H4Cl2O2
Linear Formula ClCOCH(Cl)CH3
Formula Weight 142.97
MDL Number MFCD00000647
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Fatal if inhaled.
Flammable liquid and vapor.
Contact with water liberates toxic gas.

SDS & Safety Documents