missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Chlorodocosane, TCI America™

$122.22 - $380.42

Chemical Identifiers

CAS 42217-03-8
Molecular Formula C22H45Cl
Molecular Weight (g/mol) 345.052
MDL Number MFCD00039383
InChI Key OACXFSZVCDOBKF-UHFFFAOYSA-N
Synonym Docosyl Chloride
PubChem CID 545602
IUPAC Name 1-chlorodocosane
SMILES CCCCCCCCCCCCCCCCCCCCCCCl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C07755G View Documents TCI America
C07755G
5g Each for $122.22
 
C077525G View Documents TCI America
C077525G
25g Each for $380.42
 
Specifications

Chemical Identifiers

42217-03-8
345.052
OACXFSZVCDOBKF-UHFFFAOYSA-N
545602
CCCCCCCCCCCCCCCCCCCCCCCl
C22H45Cl
MFCD00039383
Docosyl Chloride
1-chlorodocosane

Specifications

White-Yellow
5g
C22H45Cl
Docosyl Chloride
CCCCCCCCCCCCCCCCCCCCCCCl
345.052
345.05
≥98.0% (GC)
41°C
42217-03-8
MFCD00039383
OACXFSZVCDOBKF-UHFFFAOYSA-N
1-chlorodocosane
545602
Crystal-Powder at 20°C
1-Chlorodocosane
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0066&(2)-0068

TSCA : No