Learn More
1-Chlorodecane, 97-99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 108980010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1-Chlorodecane | |
1002-69-3 | |
100.0 | |
223°C | |
Authentic | |
Glass bottle | |
1.4362 to 1.4382 | |
MFCD00000956 | |
01, 168 | |
decyl chloride, decane, 1-chloro, n-decyl chloride, decane, chloro, unii-l0hsc44e5x, decyl chloride mixed isomers, chlorodecane, l0hsc44e5x, chlorodecane mixed isomers, 1-chlorodecane | |
ZTEHOZMYMCEYRM-UHFFFAOYSA-N | |
1-chlorodecane | |
13848 | |
176.73 |
97-99% | |
97.0 | |
0.8600g/mL | |
83°C | |
97% min. (GC) | |
C10H21Cl | |
CH3(CH2)9Cl | |
1L | |
0.86 | |
Solubility in water: 0.01g/L (20°C) | |
CCCCCCCCCCCl | |
176.73 | |
1.94 mPa.s (20°C) | |
97-99% |
Chemical Identifiers
1002-69-3 | |
176.73 | |
ZTEHOZMYMCEYRM-UHFFFAOYSA-N | |
13848 | |
CCCCCCCCCCCl |
C10H21Cl | |
MFCD00000956 | |
decyl chloride, decane, 1-chloro, n-decyl chloride, decane, chloro, unii-l0hsc44e5x, decyl chloride mixed isomers, chlorodecane, l0hsc44e5x, chlorodecane mixed isomers, 1-chlorodecane | |
1-chlorodecane |
Safety and Handling
GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
GHS P Statement:
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 213-691-0
TSCA : TSCA
RUO â Research Use Only