missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Chlorobutane, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A11494AE

 View more versions of this product

Catalog No. AAA11494AE



Description

Description

1-Chlorobutane is used as intermediates and solvents. It is used for HPLC, spectrophotometry and environmental testing. It is used as an alkylating reagent in organic and organometallic compound synthesis. It is also employed as an anthelmintic in veterinary medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chlorobutane is used as intermediates and solvents. It is used for HPLC, spectrophotometry and environmental testing. It is used as an alkylating reagent in organic and organometallic compound synthesis. It is also employed as an anthelmintic in veterinary medicine.

Solubility
Miscible with alcohol and ether. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications

Specifications

1-Chlorobutane
-123°C
77°C to 78°C
>99%
C4H9Cl
MFCD00001009
UN1127
14,1560
Miscible with alcohol and ether. Immiscible with water.
CCCCCl
92.57
92.57
Liquid
109-69-3
0.886
−12°C (10°F)
Pungent
1.4024
100 mL
1730909
butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane
VFWCMGCRMGJXDK-UHFFFAOYSA-N
1-chlorobutane
8005
≥99%

Chemical Identifiers

109-69-3
92.57
VFWCMGCRMGJXDK-UHFFFAOYSA-N
8005
CCCCCl
C4H9Cl
MFCD00001009
butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane
1-chlorobutane
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c

H225-H304

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CHLOROBUTANES

EINECSNumber : 203-696-6

RTECSNumber : EJ6300000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only