Please login to your online account to display your discounted pricing

1-Chloroanthraquinone, 98%, ACROS Organics™


Quantity 100g

Manufacturer: thermo fisher scientific  108601000

 View more versions of this product

Catalog No. AC108601000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Water 0.5% max.
Color Light yellow to yellow
Melting Point 157° to 162°C
Quantity 100g
Chemical Name or Material 1-Chloroanthraquinone
Name Note 98%
CAS 82-44-0
CAS Min % 97.5
CAS Max % 100.0
Physical Form Light yellow to yellow powder or chunks
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C14H7ClO2
Formula Weight 242.66g/mol
MDL Number MFCD00001189
RTECS Number CB6150000
EINECS Number 201-421-4
TSCA TSCA

Specifications

Chemical Name or Material 1-Chloroanthraquinone
Name Note 98%
CAS 82-44-0
CAS Min % 97.5
CAS Max % 100.0
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C14H7ClO2
Formula Weight 242.66g/mol
MDL Number MFCD00001189