Please login to your online account to display your discounted pricing

1-Chloroanthraquinone, 98%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  AC108600050

 View more versions of this product

Catalog No. AC108600050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder or Chunks
Assay ≥97.5%
Formula Weight 242.66
Quantity 5g
Color Light yellow to yellow
EINECS Number 201-421-4
TSCA TSCA
RTECS Number CB6150000
Melting Point 157° to 162°C
CAS Max % 100.0
Chemical Name or Material 1-Chloroanthraquinone
Packaging Glass Bottle
Name Note 98%
Assay Percent Range 98%
Molecular Formula C14H7ClO2
MDL Number MFCD00001189
CAS 82-44-0
CAS Min % 97.5
Water 0.5% max.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 1-Chloroanthraquinone
Name Note 98%
CAS 82-44-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C14H7ClO2
Formula Weight 242.66
MDL Number MFCD00001189