Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Chloroanthraquinone, 98%

$37.85 - $124.82

Chemical Identifiers

Chemical Name or Material 1-Chloroanthraquinone
CAS 82-44-0
Molecular Formula C14H7ClO2
Formula Weight 242.66
MDL Number MFCD00001189
Beilstein 1912752
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2452222

  SDS
alfa aesar™
B2452222
100g Each for $37.85

AAB2452236

  SDS
alfa aesar™
B2452236
500g Each for $124.82
Description & Specifications

Specifications

Formula Weight 242.66
Chemical Name or Material 1-Chloroanthraquinone
TSCA TSCA
Assay 98%
Molecular Formula C14H7ClO2
MDL Number MFCD00001189
CAS 82-44-0
Beilstein 1912752
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number CB6150000
Melting Point 158° to 162°C
Quantity 100g

Applications
It is used as a dyestuff intermediate.

Solubility
Soluble in hot toluene. (almost transparency)

Notes
Store at room temperature. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.