missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Chloroadamantane, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 264240500

 View more versions of this product

Catalog No. AC264240500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1-chloroadamantane
163.0°C to 166.0°C
935-56-8
100.0
C10H15Cl
1-adamantyl chloride, 1-chlorotricyclo 3.3.1.13,7 decane, 1-chlorotricyclo 3.3.1.1~3,7~ decane, 3-chloroadamantane, 3r,5s,7s-1-chloroadamantane, adamantane, 1-chloro, adamantyl chloride, tricyclo 3.3.1.13,7 decane, 1-chloro
C1C2CC3CC1CC(C2)(C3)Cl
170.68
170.68
Glass bottle
50g
97.5
97.5% min. (GC)
MFCD00075627
OZNXTQSXSHODFR-UHFFFAOYSA-N
1-chloroadamantane
64154
98%

Chemical Identifiers

935-56-8
170.68
OZNXTQSXSHODFR-UHFFFAOYSA-N
64154
C1C2CC3CC1CC(C2)(C3)Cl
C10H15Cl
MFCD00075627
1-adamantyl chloride, 1-chlorotricyclo 3.3.1.13,7 decane, 1-chlorotricyclo 3.3.1.1~3,7~ decane, 3-chloroadamantane, 3r,5s,7s-1-chloroadamantane, adamantane, 1-chloro, adamantyl chloride, tricyclo 3.3.1.13,7 decane, 1-chloro
1-chloroadamantane
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 213-305-

SDS Documents