missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Chloroacetyl-2-(trifluoroacetyl)hydrazine, 97%

Manufacturer:  Alfa Aesar™ H5464706

 View more versions of this product

Catalog No. AAH5464706



Specifications

Specifications

1-Chloroacetyl-2-(trifluoroacetyl)hydrazine
762240-99-3
MFCD07779449
DYKIVKLXFDNBMY-UHFFFAOYSA-N
N'-(2-chloroacetyl)-2,2,2-trifluoroacetohydrazide
11961370
97%
5g
C4H4ClF3N2O2
1-chloroacetyl-2-trifluoroacetyl hydrazine
C(C(=O)NNC(=O)C(F)(F)F)Cl
204.533
204.53

Chemical Identifiers

762240-99-3
204.533
DYKIVKLXFDNBMY-UHFFFAOYSA-N
11961370
C(C(=O)NNC(=O)C(F)(F)F)Cl
C4H4ClF3N2O2
MFCD07779449
1-chloroacetyl-2-trifluoroacetyl hydrazine
N'-(2-chloroacetyl)-2,2,2-trifluoroacetohydrazide
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents