Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Chloro-8-phenyloctane, 97%

$94.47 - $366.15

Chemical Identifiers

Chemical Name or Material 1-Chloro-8-phenyloctane
CAS 61440-32-2
Molecular Formula C6H5(CH2)8Cl
Formula Weight 224.78
MDL Number MFCD00039394
Beilstein 6289454
Synonym 8-Phenyl-n-octyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0901703

  SDS
alfa aesar™
L0901703
1g Each for $94.47

AAL0901706

  SDS
alfa aesar™
L0901706
5g Each for $366.15
Description & Specifications

Specifications

Formula Weight 224.78
Chemical Name or Material 1-Chloro-8-phenyloctane
CAS 61440-32-2
Molecular Formula C6H5(CH2)8Cl
MDL Number MFCD00039394
Synonym 8-Phenyl-n-octyl chloride
Beilstein 6289454
Quantity 1g
Boiling Point 108° to 110°C (0.4mmHg)
Flash Point >110°C (230°F)
Assay 97%