Please login to your online account to display your discounted pricing

1-Chloro-8-hydroxyquinoline 95%, ACROS Organics™


Quantity 500g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  109305000

 View more versions of this product

Catalog No. AC109305000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Assay ≥94%
Formula Weight 179.61
Quantity 500g
Color Light green to gray
Beilstein 21, 95
Merck Index 15, 2405
EINECS Number 204-978-1
TSCA TSCA
RTECS Number VC4590000
Melting Point 121° to 126°C
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material 5-Chloro-8-hydroxyquinoline
Infrared Spectrum Authentic
Synonym 5-Chloro-8-quinolinol
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Packaging Plastic bottle
Name Note 95%
Assay Percent Range 95%
Molecular Formula C9H6ClNO
MDL Number MFCD00006788
CAS 130-16-5
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 5-Chloro-8-hydroxyquinoline
Name Note 95%
CAS 130-16-5
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C9H6ClNO
Formula Weight 179.61
MDL Number MFCD00006788
Synonym 5-Chloro-8-quinolinol
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents