Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Chloro-6-phenylhexane, 98+%

$146.16 - $146.16

Chemical Identifiers

Chemical Name or Material 1-Chloro-6-phenylhexane
CAS 71434-68-9
Molecular Formula C6H5(CH2)6Cl
Formula Weight 196.72
MDL Number MFCD00013698
Beilstein 2088276
Synonym 6-Phenyl-n-hexyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0894903

  SDS
alfa aesar™
L0894903
1g Each for $146.16
Description & Specifications

Specifications

Formula Weight 196.72
CAS 71434-68-9
Molecular Formula C6H5(CH2)6Cl
Chemical Name or Material 1-Chloro-6-phenylhexane
MDL Number MFCD00013698
Beilstein 2088276
Flash Point >110°C (230°F)
Boiling Point 88° to 91°C (0.4mmHg)
Synonym 6-Phenyl-n-hexyl chloride
Quantity 1g
Assay ≥98%