Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Chloro-6,6-dimethyl-2-hepten-4-yne, cis + trans, tech. 90%

$128.82 - $128.82

Chemical Identifiers

Chemical Name or Material 1-Chloro-6,6-dimethyl-2-hepten-4-yne
CAS 126764-17-8
Name Note cis + trans
Purity Grade Technical Grade
Molecular Formula C9H13Cl
Formula Weight 156.65
MDL Number MFCD04039160
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2532006

  SDS
alfa aesar™
B2532006
5g Each for $128.82
Description & Specifications

Specifications

Formula Weight 156.65
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Name Note cis + trans
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 126764-17-8
Molecular Formula C9H13Cl
Chemical Name or Material 1-Chloro-6,6-dimethyl-2-hepten-4-yne
Health Hazard 1 Warning
Quantity 5g
Purity Grade Technical Grade
MDL Number MFCD04039160
Assay 90%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.